missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TER-119 Monoclonal Antibody (TER119), PE
GREENER_CHOICE

Rat Monoclonal Antibody

€ 225.00 - € 506.00

Specifications

Antigen TER-119
Clone TER119
Concentration 0.1 mg/mL
Content And Storage 4°C, do not freeze
Applications Flow Cytometry
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15324101
GREENER_CHOICE
View Documents
Invitrogen™
MA517823
50 μg
€ 225.00
50µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15334101
View Documents
Invitrogen™
MA517824
300 μg
€ 506.00
300µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TER-119 Monoclonal Antibody for Flow

TER-119 is a lineage marker for erythroid cells from early proerythroblast to mature erythrocyte stages in adult blood, spleen, and bone marrow. It is also present in yolk sac, and fetal and newborn liver. The TER-119 antigen is not expressed by cells of earlier erythroid development at BFU-e (blast-forming unit erythroid) stage or CFU-e (colony-forming unit erythroid) stage.
Specifications

Specifications

TER-119
0.1 mg/mL
Flow Cytometry
PE
Rat
RUO
PBS with 4-5mg/mL BSA and 0.02% sodium azide
0
104231
Day 14 BALB/c fetal liver cells.
Antibody
TER119
4°C, do not freeze
Monoclonal
Liquid
IgG2b
Mouse
Ly76
Ly76; Ly-76; TER119
Ly76
Primary
Protein G
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.