Learn More
TER-119 Rat anti-Mouse, APC, Clone: TER-119, Proteintech
Rat Monoclonal Antibody
Brand: Proteintech APC-65149-100UG
Description
The TER-119 antibody from Proteintech is a rat monoclonal antibody generated with day-14 fetal liver cells from a c57bl/6 mouse of mouse TER-119. This antibody recognizes mouse antigen. The TER-119 antibody has been validated for the following applications: FC analysis. This antibody is apc.
Specifications
| TER-119 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Rat | |
| Primary | |
| Mouse |
| Flow Cytometry | |
| TER-119 | |
| APC | |
| 100 μg | |
| 104231 | |
| IgG2b |