Learn More
TER-119 Rat anti-Mouse, APC, Clone: TER-119, Proteintech
Rat Monoclonal Antibody
Brand:  Proteintech APC-65149-100UG
Product Code. 16830526
  • € 76.00 / 100µg
Description
The TER-119 antibody from Proteintech is a rat monoclonal antibody generated with day-14 fetal liver cells from a c57bl/6 mouse of mouse TER-119. This antibody recognizes mouse antigen. The TER-119 antibody has been validated for the following applications: FC analysis. This antibody is apc.
Specifications
Specifications
TER-119
Monoclonal
0.2 mg/mL
Rat
Primary
Mouse
Flow Cytometry
TER-119
APC
100 μg
104231
IgG2b