missing translation for 'onlineSavingsMsg'
Learn More

Terbinafine, MedChemExpress

Product Code. 16411256
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
200 mg
Unit Size:
100mg
200mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16411256 200 mg 200mg
16497276 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16411256 Supplier Medchem Express Supplier No. HY17395A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Terbinafine (TDT 067) is an orally active and potent antifungal agent. Terbinafine is a potent non-competitive inhibitor of squalene epoxidase from Candida, with a Ki of 30 nM. Terbinafine also shows antibacterial activity against certain Gram-positive and Gram-negative bacteria.

Terbinafine (TDT 067) is an orally active and potent antifungal agent. Terbinafine is a potent non-competitive inhibitor of squalene epoxidase from Candida, with a Ki of 30 nM. Terbinafine also shows antibacterial activity against certain Gram-positive and Gram-negative bacteria.

Specifications

Chemical Name or Material Terbinafine
Physical Form Solid
CAS 91161-71-6
Color White
Quantity 200 mg
Purity Grade Notes Research
Molecular Formula C21H25N
For Use With (Application) COVID-19-immunoregulation
Synonym TDT 067
Solubility Information DMSO : ≥ 100 mg/mL (343.14 mM)
SMILES CN(C/C=C/C#CC(C)(C)C)CC1=C2C=CC=CC2=CC=C1
Molecular Weight (g/mol) 291.43
Formula Weight 291.43
Percent Purity 98.83%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.