missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TERF2IP Antibody (78B356.1), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-56526AF405
This item is not returnable.
View return policy
Description
TERF2IP Monoclonal specifically detects TERF2IP in Human samples. It is validated for Western Blot.
Specifications
| TERF2IP | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| dopamine receptor interacting protein 5, Dopamine receptor-interacting protein 5, DRIP5, hRap1, RAP1 homolog, RAP1TERF2-interacting telomeric protein 1, Repressor/activator protein 1 homolog, telomeric repeat binding factor 2, interacting protein, telomeric repeat-binding factor 2-interacting protein 1, TRF2-interacting telomeric protein 1, TRF2-interacting telomeric RAP1 protein | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| 78B356.1 | |
| Western Blot | |
| Q9NYB0 | |
| TERF2IP | |
| A synthetic peptide corresponding amino acids 212-235 of human Rap1 was used to immunize mice. | |
| 0.1 mL | |
| Cellular Markers, Chromatin Research, DNA Repair, Endosome Markers, Lysosome Markers | |
| 54386 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction