missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TERF2IP Antibody (RAP1 5G7/1), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB120-14403H
This item is not returnable.
View return policy
Description
TERF2IP Monoclonal antibody specifically detects TERF2IP in Human samples. It is validated for Western Blot, ELISA
Specifications
| TERF2IP | |
| Monoclonal | |
| HRP | |
| dopamine receptor interacting protein 5, Dopamine receptor-interacting protein 5, DRIP5, hRap1, RAP1 homolog, RAP1TERF2-interacting telomeric protein 1, Repressor/activator protein 1 homolog, telomeric repeat binding factor 2, interacting protein, telomeric repeat-binding factor 2-interacting protein 1, TRF2-interacting telomeric protein 1, TRF2-interacting telomeric RAP1 protein | |
| Human TERF2IP | |
| 0.1 mL | |
| Cellular Markers, Chromatin Research, DNA Repair, Endosome Markers, Lysosome Markers | |
| 54386 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, ELISA | |
| RAP1 5G7/1 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction