missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ternidazole, 95%, Thermo Scientific Chemicals

Product Code. 15753457 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15753457 1 g 1g
15763457 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15753457 Supplier Thermo Scientific Chemicals Supplier No. 461950010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1077-93-6
Molecular Formula C7H11N3O3
Molecular Weight (g/mol) 185.18
InChI Key DUOHVNSMLSPTMI-UHFFFAOYSA-N
Synonym ternidazole, unii-4n8r018qb0, 1-3-hydroxypropyl-2-methyl-5-nitro-1h-imidazole, 3-2-methyl-5-nitroimidazol-1-yl propan-1-ol, ternidazol, ternidazolum, ternidazole inn, ternidazol inn-spanish, ternidazolum inn-latin
PubChem CID 68944
IUPAC Name 3-(2-methyl-5-nitroimidazol-1-yl)propan-1-ol
SMILES CC1=NC=C(N1CCCO)[N+](=O)[O-]

Specifications

Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 185.18
Percent Purity 97%
Chemical Name or Material Ternidazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.