Learn More
MP Biomedicals™ Terrific Broth II, Dehydrated Media
Product Code. 11357699 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1000 g
5000 g
Unit Size:
1kg
5kg
Product Code. 11357699
Brand: MP Biomedicals™ 113046031
Rich medium for the growth of E. coli and certified for plasmid and other bacterial cultures in liquid medium
  • Proprietary formulation
  • Use 50 capsules per liter
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Molecular biology Certified™ FDA
Content And Storage 15°C to 30°C
Format Capsule
Grade Molecular biology
Product Type Microbiology Media
Formulation Yeast extract, Tryptone broth, Sodium Chloride and Vitamins
For Use With (Application) For growth of Escherichia coli and certified for plasmid and other bacterial cultures in liquid medium
Quantity 1000 g