missing translation for 'onlineSavingsMsg'
Learn More

Melford Terrific Broth, Powder

Product Code. 30018821
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
Unit Size:
1kg
500g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018821 500 g 500g
30018820 1 kg 1kg
30018822 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30018821 Supplier Melford Supplier No. T15000500.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Enriched media for maintenance and propogation of E.

Enriched media for maintenance and propogation of E. coli and plasmid growth. Produces higher cell densities resulting in greater yields of DNA or recombinant problem. Glycerol is used as a carbon source with this formulation instead of glucose as it will not ferment to acetic acid.

Ingredients:

Casein Digest Peptone: 12 g/l

Yeast Extract: 24 g/l

Dipotassium phosphate: 9.4 g/l

Monopotassium phosphate: 2.2 g/l

Specifications

Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.