Learn More
tert-Butyl (3-oxocyclobutyl)carbamate, 95%, Thermo Scientific Chemicals
Catalog Number p-4380062
Click to view available options
Quantity:
1 g
5 g
154748-49-9
C9H15NO3
185.22
FNHPTFKSPUTESA-UHFFFAOYSA-N
tert-butyl 3-oxocyclobutylcarbamate, tert-butyl 3-oxocyclobutyl carbamate, tert-butyl n-3-oxocyclobutyl carbamate, 3-oxo-cyclobutyl-carbamic acid tert-butyl ester, 3-boc-amino cyclobutanone, 2-methyl-2-propanyl 3-oxocyclobutyl carbamate, 3-tert-butyloxycarbonylamino cyclobutan-1-one, 3-oxocyclobutylcarbamic acid, 1,1-dimethylethyl ester, carbamic acid, 3-oxocyclobutyl-, 1,1-dimethylethyl ester, carbamic acid, n-3-oxocyclobutyl-, 1,1-dimethylethyl ester
23374584
tert-butyl N-(3-oxocyclobutyl)carbamate
CC(C)(C)OC(=O)NC1CC(=O)C1
154748-49-9
C9H15NO3
185.22
FNHPTFKSPUTESA-UHFFFAOYSA-N
tert-butyl 3-oxocyclobutylcarbamate, tert-butyl 3-oxocyclobutyl carbamate, tert-butyl n-3-oxocyclobutyl carbamate, 3-oxo-cyclobutyl-carbamic acid tert-butyl ester, 3-boc-amino cyclobutanone, 2-methyl-2-propanyl 3-oxocyclobutyl carbamate, 3-tert-butyloxycarbonylamino cyclobutan-1-one, 3-oxocyclobutylcarbamic acid, 1,1-dimethylethyl ester, carbamic acid, 3-oxocyclobutyl-, 1,1-dimethylethyl ester, carbamic acid, n-3-oxocyclobutyl-, 1,1-dimethylethyl ester
23374584
tert-butyl N-(3-oxocyclobutyl)carbamate
CC(C)(C)OC(=O)NC1CC(=O)C1
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 122°C to 124°C
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Formula Weight 185.22
Percent Purity 95%
Quantity 1 g
Chemical Name or Material tert-Butyl (3-oxocyclobutyl)carbamate
compliance-icons
Product Identifier
  • tert-Butyl (3-oxocyclobutyl)carbamate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.