Learn More
tert-Butyl Carbazate, TRC
Code produit 30340975
Modifier l'affichage
Click to view available options
Quantity:
50 g
Conditionnement:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantity Conditionnement
30340975 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 30340975 Fournisseur Toronto Research Chemicals Code fournisseur TRCB69217050G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Spécification
Chemical Name or Material tert-Butyl Carbazate
CAS 870-46-2
Molecular Formula C5H12N2O2
Quantity 50 g
Synonym tert-Butyl Ester Carbazic Acid, (tert-Butoxycarbonyl)hydrazide, (tert-Butoxycarbonyl)hydrazine, (tert-Butyloxycarbonyl)hydrazide, 1,1-Dimethylethyl Carbazate, 1,1-Dimethylethyl Hydrazinecarboxylate, 1-(tert-Butoxycarbonyl)hydrazine, Boc-hydrazine, Hydrazinecarboxylic Acid tert-Butyl Ester, N-(tert-Butoxycarbonyl)hydrazine, N-Boc-Hydrazine, NSC 60250, tert-Butyl N-aminocarbamate, tert-Butyl Carbazate, tert-Butyl Hydrazinecarboxylate, tert-Butyl Hydrazinoformate
InChI Formula InChI=1S/C5H12N2O2/c1-5(2,3)9-4(8)7-6/h6H2,1-3H3,(H,7,8)
SMILES CC(C)(C)OC(=O)NN
IUPAC Name tert-butyl N-aminocarbamate
Molecular Weight (g/mol) 132.16
Formula Weight 132.0899