Learn More
tert-Butyl N-[2-[2-(6-chlorohexyloxy)ethoxy]ethyl]carbamate, TRC
Specifications
Specifications
| Chemical Name or Material | tert-Butyl N-[2-[2-(6-chlorohexyloxy)ethoxy]ethyl]carbamate |
| CAS | 920264-35-3 |
| Molecular Formula | C15H30ClNO4 |
| Quantity | 10 mg |
| Synonym | N-[2-[2-[(6-Chlorohexyl)oxy]ethoxy]ethyl]-carbamic Acid 1,1-Dimethylethyl Ester |
| InChI Formula | InChI=1S/C15H30ClNO4/c1-15(2,3)21-14(18)17-9-11-20-13-12-19-10-7-5-4-6-8-16/h4-13H2,1-3H3,(H,17,18) |
| SMILES | CC(C)(C)OC(NCCOCCOCCCCCCCl)=O |
| IUPAC Name | tert-butyl N-[2-[2-(6-chlorohexoxy)ethoxy]ethyl]carbamate |
| Molecular Weight (g/mol) | 323.86 |
| Formula Weight | 323.1863 |