Learn More
tert-Butyl vinyl ether, 98%, stab. with ca 0.1% N,N-diethylaniline
Product Code. p-7030604
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 15427785
Brand: Thermo Scientific Alfa Aesar H30770.06
Request Bulk or Custom Format
CAS: 926-02-3 | C6H12O | 100.16 g/mol
tert-Butyl vinyl ether is used in the synthesis of 3-tert-Butoxycyclobutanone by reacting with ketene using zinc(II) chloride as a catalyst. It acts as an intermediate for the functional vinyl monomer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
tert-Butyl vinyl ether is used in the synthesis of 3-tert-Butoxycyclobutanone by reacting with ketene using zinc(II) chloride as a catalyst. It acts as an intermediate for the functional vinyl monomer.
Solubility
Miscible with alcohol, carbon disulfide, chloroform and diethyl ether. Slightly miscible with water.
Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 926-02-3
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD00048246
InChI Key PGYJSURPYAAOMM-UHFFFAOYSA-N
Synonym tert-butyl vinyl ether, 2-ethenyloxy-2-methylpropane, propane, 2-ethenyloxy-2-methyl, propane,2-ethenyloxy-2-methyl, t-butyl vinyl ether, tert-butylvinyl ether, vinyltertiary-butylether, tertiarybutyl vinyl ether, vinyl tert.-butyl ether, tertiary butyl vinyl ether
PubChem CID 70220
IUPAC Name 2-ethenoxy-2-methylpropane
SMILES CC(C)(C)OC=C
Specifications
Density 0.762
Boiling Point 75°C to 76°C
Flash Point −17°C (1°F)
Odor Characteristic
Linear Formula (CH3)3COCH=CH2
Refractive Index 1.399
Quantity 5 g
UN Number UN3271
Solubility Information Miscible with alcohol,carbon disulfide,chloroform and diethyl ether. Slightly miscible with water.
Formula Weight 100.16
Percent Purity 98%
Chemical Name or Material tert-Butyl vinyl ether, stabilized with ≈0.1% N,N-diethylaniline
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • tert-Butyl vinyl ether, 98%, stab. with ca 0.1% N,N-diethylaniline
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : tert-Butyl vinyl ether, N,N-diethylaniline