Learn More
tert-Butyldimethylsilanol, TRC
Specifications
Specifications
| Chemical Name or Material | tert-Butyldimethylsilanol |
| CAS | 18173-64-3 |
| Molecular Formula | C6H16OSi |
| Quantity | 2.5 mg |
| InChI Formula | InChI=1S/C6H16OSi/c1-6(2,3)8(4,5)7/h7H,1-5H3 |
| SMILES | CC(C)(C)[Si](C)(C)O |
| IUPAC Name | tert-butyl-hydroxy-dimethylsilane |
| Molecular Weight (g/mol) | 132.28 |
| Formula Weight | 132.097 |