missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TESC Polyclonal Antibody

Description
Immunogen sequence: YRNVVEELLS GNPHIEKESA RSIADGAMME AASVCMGQME PDQVYEGITF EDFLKIWQGI DIETKMHVRF LNMETMALCH; Positive Samples: HeLa, U87, 293T; Cellular Location: Cell membrane, Cell projection, Cytoplasm, Lipid-anchor, Membrane, Nucleus, lamellipodium, ruffle membrane.
Essential for the coupling of ERK cascade activation with the expression of ETS family genes in megakaryocytic differentiation.
Specifications
Specifications
| Antigen | TESC |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.65 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | TESC |
| Gene Accession No. | Q96BS2 |
| Gene Alias | 1010001A17Rik; 2410011K10Rik; Calcineurin B homologous protein 3; calcineurin-like EF hand protein 3; CHP3; RGD1566317; TE-1; Tesc; Tescalcin; TSC |
| Gene Symbols | TESC |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?