Learn More
Hach™ Test 'N Tube (TNT) Cuvette Tests
Product Code. 15192923
Click to view available options
For Use With (Application):
COD Determination
Nitrite Determination
Total Phosphate Determination
Unit Size:
Each
Pack of 25
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 15192923
Brand: Hach™ 2125951
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Provides a self-contained package for mixing and measurement. Hach™ Test 'N Tube (TNT) Hach™ Test 'N Tube (TNT) Cuvette Tests include 16mm capped tubes and all necessary reagents in one convenient package for reliable results and easy handling.
Provides a self-contained package for mixing and measurement. Hach™ Test 'N Tube (TNT) Hach™ Test 'N Tube (TNT) Cuvette Tests include 16mm capped tubes and all necessary reagents in one convenient package for reliable results and easy handling.
Test 'N Tube products enable convenient testing with ready to use ampules and pre-measured reagents all packed into one convenient package. Available for determinations of a variety of parameters.
  • Test 'N Tube: Capped 16mm vials provide a safe, self-contained package for mixing and measurement
  • Powder Pillows: Premeasured, single dose packets of powdered reagent
  • Dropper bottles: For easy addition of liquid reagents
  • Precisely dosed reagents and test cuvettes for easy planning and inventory control
Specifications
Description Colorimetric determination cuvette tests
For Use With (Application) COD Determination
For Use With (Equipment) Hach DR900
Includes 16mm Tubes of Digestion Solution
No. of Tests 25 (1 for blank needed)
Quantity 1 Kit
Range 20 to 1500 mg/L O2
Test Method Dichromate
Type Test 'N Tube
compliance-icons compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Carcinogenicity Category 1A
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Germ cell mutagenicity Category 1B
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
  • EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.