Learn More
Reitenspiess™ Test Tube Brush
Product Code. 10403131
Change view
Click to view available options
Length (Metric):
190 mm
Diameter (Metric) Bristle:
10 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Bristle unitSize
10403131 190 mm 10 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10403131 Supplier Reitenspiess™ Supplier No. 12200334
Only null left
Add to basket
Add to basket
Use a Reitenspiess™ Test Tube Brush for cleaning, disinfecting and sterilizing test tubes as necessary in the laboratory. .
  • Natural bristle
  • Various lengths available
  • Various diameters
Specifications
Product Type Test Tube Brush
Diameter (Metric) Bristle 10 mm
Length (Metric) 190 mm
Quantity 1 Ea.