Learn More
Reitenspiess™ Test Tube Brush
Product Code. p-8008528
Click to view available options
Length (Metric):
190 mm
Diameter (Metric) Bristle:
10 mm
Unit Size:
Each
Product Code. 10403131
Brand: Reitenspiess™ 12200334
Only null left
Add to basket
Edge
Add to basket
Use a Reitenspiess™ Test Tube Brush for cleaning, disinfecting and sterilizing test tubes as necessary in the laboratory. .
  • Natural bristle
  • Various lengths available
  • Various diameters
Specifications
Product Type Test Tube Brush
Diameter (Metric) Bristle 10 mm
Length (Metric) 190 mm
Quantity 1 Ea.