Learn More
Usbeck Laborgeraete™ Test Tube Holder
Product Code. 10771653
Change view
Click to view available options
Length (Metric):
130 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10771653 130 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10771653 Supplier Usbeck Laborgeraete™ Supplier No. 5570
  • ONLINE EXCLUSIVE PRICE
    € 4.30
    € 4.25 / Pack of 1
    Save € 0.05
    1% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Product Type Test Tube Holder
For Use With (Application) Test Tube
Length (Metric) 130 mm
Material Stainless Steel