Learn More
Julabo™ Test Tube Racks for Open Heating Bath Circulator
Click to view available options
Material:
Polypropylene
Stainless Steel
Holds:
21 Test Tubes, 30/31 mm dia.
90 Microliter Tubes, 11/12 mm dia.
90 Test Tubes, 12/13 mm dia.
Unit Size:
Each
Specifications
Specifications
| Description | Test tube rack |
| For Use With (Equipment) | SW22, SW23, TW12, TW20, TW8 shaking water bath |
| Holds | 21 Test Tubes, 30/31 mm dia. |
| Material | Stainless Steel |