missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasintrumente Test Tubes with NS Joint Without Graduation

Product Code. 11556944
Click to view available options
Diameter (Metric):
18 mm
22 mm
28 mm
Height (Metric):
150 mm
200 mm
Closure Type:
Not Included
Stopper
Unit Size:
Each
Pack of 10
This item is not returnable. View return policy

Product Code. 11556944

Brand: Lenz Laborglasintrumente 05700038

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.

  • With round bottom
  • With NS socket
  • Available without stopper
  • With PE stopper or with hexagonal hollow glass stopper
  • Made of DURAN™ tubing

Specifications

Material Borosilicate Glass (3.3)
Height (Metric) 150 mm
Diameter (Metric) 28 mm
Closure Type Not Included
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.