missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Testican 1/SPOCK1 Antibody [DyLight 405], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-33162V
This item is not returnable.
View return policy
Description
Testican 1/SPOCK1 Polyclonal antibody specifically detects Testican 1/SPOCK1 in Human samples. It is validated for ELISA,Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| Testican 1/SPOCK1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 6695 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 405 | |
| FLJ37170, Protein SPOCK, sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 1, SPOCKcwcv and kazal-like domains proteoglycan (testican) | |
| Produced in rabbits immunized with purified, recombinant Human Testican 1/SPOCK1 (Uniprot#: Q08629; Met1-Trp439) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction