missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Testis expressed 264 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
€ 369.00 - € 554.00
Specifications
| Antigen | Testis expressed 264 |
|---|---|
| Dilution | Western Blot -Reactivity reported in (PMID: 31006538)., Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18434342
|
Novus Biologicals
NBP1-89866-25ul |
25 μL |
€ 369.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18065075
|
Novus Biologicals
NBP1-89866 |
0.1 mL |
€ 554.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Testis expressed 264 Polyclonal specifically detects Testis expressed 264 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Testis expressed 264 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 51368 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:AIDTQVDGTGADTMSDTSSVSLEVSPGSRETSAATLSPGASSRGWDDGDTRSEHSYSESGASGSSFEELDLEGEGPLGESRLDPGTEPLGTTKWLWEPTAPEKGKE | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot -Reactivity reported in (PMID: 31006538)., Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| DKFZp451H0417, Putative secreted protein Zsig11, SIG11, testis expressed 264, testis expressed gene 264, testis expressed sequence 264, testis-expressed sequence 264 protein, ZSIG11FLJ13935 | |
| TEX264 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title