missing translation for 'onlineSavingsMsg'
Learn More

Testosterone 17-Hexahydrobenzoate, TRC

Product Code. 30418501
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30418501 100 mg 100mg
30421865 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30418501 Supplier Toronto Research Chemicals Supplier No. TRCT155090100MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Testosterone 17-Hexahydrobenzoate
CAS 14191-92-5
Molecular Formula C26 H38 O3
Quantity 100 mg
Synonym Sterandryl-A, Tardosterandryl-Depot, Testodur, Testosterone 17-Hexahydrobenzoate, Testosterone Cyclohexanoate, Testosterone Hexahydrobenzoate, Virex-Retard, Benzotest, Durandron, Sterandryl Retard, Cyclohexanecarboxylate Testosterone, Cyclohexanecarboxylic Acid Ester with Testosterone
InChI Formula InChI=1S/C26H38O3/c1-25-14-12-19(27)16-18(25)8-9-20-21-10-11-23(26(21,2)15-13-22(20)25)29-24(28)17-6-4-3-5-7-17/h16-17,20-23H,3-15H2,1-2H3/t20?,21?,22?,23-,25-,26-/m0/s1
SMILES C[C@]12CCC3C(CCC4=CC(=O)CC[C@]34C)C1CC[C@@H]2OC(=O)C5CCCCC5
IUPAC Name [(10R,13S,17S)-10,13-dimethyl-3-oxo-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl] cyclohexanecarboxylate
Molecular Weight (g/mol) 398.58
Formula Weight 398.2821
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.