missing translation for 'onlineSavingsMsg'
Learn More

Testosterone Cypionate-d3, TRC

Product Code. 30416246
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30416246 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30416246 Supplier Toronto Research Chemicals Supplier No. TRCT1551021MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material Testosterone Cypionate-d3
CAS 876054-56-7
Molecular Formula C27 D3 H37 O3
Quantity 1 mg
Synonym Androst-4-en-3-one-16,16,17-d3','17-(3-cyclopentyl-1-oxopropoxy)-','(17β)- (9CI), (17β)-17-(3-Cyclopentyl-1-oxopropoxy)androst-4-en-3-one-16,16,17-d3, Testosterone-16,16,17-d3 17-cyclopentanepropionate, 17β-Hydroxyandrost-4-en-3-one-16,16,17-d3 cyclopentylpropionate, Testosterone Cypionate-d3, Testosterone Cypionate-16,17,17-D3
InChI Formula InChI=1S/C27H40O3/c1-26-15-13-20(28)17-19(26)8-9-21-22-10-11-24(27(22,2)16-14-23(21)26)30-25(29)12-7-18-5-3-4-6-18/h17-18,21-24H,3-16H2,1-2H3/t21-,22-,23-,24-,26-,27-/m0/s1/i11D2,24D
SMILES [2H]C1([2H])C[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]2(C)[C@@]1([2H])OC(=O)CCC5CCCC5
IUPAC Name [(8R,9S,10R,13S,14S,17S)-16,16,17-trideuterio-10,13-dimethyl-3-oxo-1,2,6,7,8,9,11,12,14,15-decahydrocyclopenta[a]phenanthren-17-yl] 3-cyclopentylpropanoate
Molecular Weight (g/mol) 415.623
Formula Weight 415.317
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.