missing translation for 'onlineSavingsMsg'
Learn More

Testosterone Hexanoate, TRC

Product Code. 30423022
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30423022 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30423022 Supplier Toronto Research Chemicals Supplier No. TRCT15512525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Testosterone caproate
CAS 10312-45-5
Molecular Formula C25 H38 O3
Quantity 25 mg
Synonym 3-Oxoandrost-4-en-17beta-yl Hexanoate, Testosterone Enantate Imp. B (EP), Testosterone Isocaproate Imp. E (EP), Androst-4-en-3-one','17-[(1-oxohexyl)oxy]-','(17β)-, Testosterone','hexanoate (6CI,7CI,8CI), (17β)-17-[(1-Oxohexyl)oxy]androst-4-en-3-one, 17β-Hydroxyandrost-4-en-3-one hexanoate, Testosterone 17-caproate, Testosterone caproate
InChI Formula InChI=1S/C25H38O3/c1-4-5-6-7-23(27)28-22-11-10-20-19-9-8-17-16-18(26)12-14-24(17,2)21(19)13-15-25(20,22)3/h16,19-22H,4-15H2,1-3H3/t19-,20-,21-,22-,24-,25-/m0/s1
SMILES CCCCCC(=O)O[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C
IUPAC Name [(8R,9S,10R,13S,14S,17S)-10,13-dimethyl-3-oxo-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl] hexanoate
Molecular Weight (g/mol) 386.57
Formula Weight 386.2821
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.