missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Testosterone High Sensitivity Competitive ELISA Kit

Product Code. 18001663
Click to view available options
Quantity:
96 Tests
Unit Size:
Pack of 96
This item is not returnable. View return policy

Product Code. 18001663

Brand: Invitrogen™ EIATES

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Testosterone is a steroid hormone from the androgen group and is found in mammals, reptiles, birds, and other vertebrates. In mammals, testosterone is primarily secreted in the testes of males and the ovaries of females, although small amounts are also secreted by the adrenal glands. It is the principal male sex hormone and an anabolic steroid.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 7.8 to 1000 pg/mL
Assay Sensitivity 2.97 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Fecal Extract, Plasma, Serum, Urine
For Use With (Equipment) Colorimetric Microplate Reader
Interassay CV <10%
Intraassay CV <6
Kit Contents Pre-coated 96 well plate, Standard, Antibody, HRP-Conjugated Detection Antibody, Assay Buffer Concentrate, Wash Buffer, TMB Substrate, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Fecal Extract 50 μL, Plasma 50 μL, Serum 50 μL, Urine 50 μL
Storage Requirements -20°C
Target Testosterone
Test Time 1 hrs 20 mins
Total Assay Time 2 hrs 30 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.