missing translation for 'onlineSavingsMsg'
Learn More

Testosterone Mouse, Clone: 4E1G2, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio MSM1-250-P0

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15931338

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

Testosterone
Monoclonal
200μg/mL
10mM PBS with 0.05% BSA and 0.05% Azide
Secreted and cytoplasmic
20 μg
Primary
IgG1 κ
Immunohistochemistry
4E1G2
Unconjugated
Mouse
Protein A/G Purified
RUO
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers