missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Tet Repressor Polyclonal Antibody
GREENER_CHOICE

Product Code. 11598890
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11598890 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11598890 Supplier Invitrogen™ Supplier No. PA14501

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PA1-4501 detects Tet repressor in bacteria samples. PA1-4501 has been successfully used in Western blot and ELISA procedures. The PA1-4501 immunogen is Residues 31-49 [AQKLGVEQPTLYWHVKNKR] of the 22 kDa Tet repressor protein. Maintain at 2-8ºC of 3 months or at -20ºC for longer periods. Avoid repeated freeze/thaw cycles.

The Tet repressor protein (TetR) regulates transcription of a family of tetracycline resistance determinants in Gram-negative bacteria. The Tet system is the most widely used regulatory system for conditional gene expression in eukaryotic cells. TetR can be used for that purpose in some organisms without further modifications. In mammals and in a large variety of other organisms, however, eukaryotic transcriptional activator or repressor domains are fused to TetR to turn it into an efficient regulator.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tet Repressor
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Alias pSC101 tetR; TetR
Host Species Rabbit
Immunogen Residues 31-49 [AQKLGVEQPTLYWHVKNKR] of the 22 kDa Tet repressor protein.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.