missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TET1 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-78966AF594
This item is not returnable.
View return policy
Description
TET1 Polyclonal specifically detects TET1 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| TET1 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| bA119F7.1, CXXC finger 6, CXXC6, CXXC6FLJ10839, FLJ41442, KIAA1676CXXC-type zinc finger protein 6, LCX, LCXEC 1.14.11.n2, Leukemia-associated protein with a CXXC domain, methylcytosine dioxygenase TET1, Ten-eleven translocation 1 gene protein, ten-eleven translocation-1, tet oncogene 1 | |
| Rabbit | |
| Affinity Purified | |
| Primary | |
| Mouse | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| TET1 | |
| A synthetic peptide made to an internal portion of the human TET1 protein (between residues 1700 to 1800). [UniProt Q8NFU7] | |
| 0.1 mL | |
| 80312.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction