missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TET1 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-97605CL1
This item is not returnable.
View return policy
Description
TET1 Polyclonal antibody specifically detects TET1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence
Specifications
| TET1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| bA119F7.1, CXXC finger 6, CXXC6, CXXC6FLJ10839, FLJ41442, KIAA1676CXXC-type zinc finger protein 6, LCX, LCXEC 1.14.11.n2, Leukemia-associated protein with a CXXC domain, methylcytosine dioxygenase TET1, Ten-eleven translocation 1 gene protein, ten-eleven translocation-1, tet oncogene 1 | |
| Produced in rabbits immunized with E. coli-derived Human TET1 fragment. | |
| 0.1 mL | |
| Cancer, Epigenetics, Neuroscience | |
| 80312 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction