missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TET2 Antibody (CL6873), Novus Biologicals™
Mouse Monoclonal Antibody
€ 369.00 - € 504.00
Specifications
| Antigen | TET2 |
|---|---|
| Clone | CL6873 |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18645951
|
Novus Biologicals
NBP2-76510-25ul |
25 μL |
€ 369.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18330621
|
Novus Biologicals
NBP2-76510 |
100 μL |
€ 504.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TET2 Monoclonal specifically detects TET2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| TET2 | |
| Monoclonal | |
| Purified | |
| Human | |
| FLJ20032, MGC125715, tet methylcytosine dioxygenase 2 | |
| TET2 | |
| IgG2b | |
| Protein A purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| CL6873 | |
| Unconjugated | |
| Mouse | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% sodium azide | |
| 54790.0 | |
| This antibody was developed against a synthetic peptide corresponding to amino acids: PDQGGSHTQTPPQKDTQKH | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title