Learn More
TET2 Antibody (CL6873) [PerCP], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-76510PCP
Product Code. 30299623
  • € 737.00 / 0.10mL
Description
TET2 Monoclonal antibody specifically detects TET2 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TET2
Monoclonal
PerCP
FLJ20032, MGC125715, tet methylcytosine dioxygenase 2
This antibody was developed using a synthetic peptide derived from Q6N021, with the exact immunogen sequence remaining proprietary.
0.1 mL
Primary
Human
Purified
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL6873
PBS
Mouse
Protein A purified
RUO
54790
Store at 4⪚C in the dark.
IgG2b