missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TET3 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98069F
This item is not returnable.
View return policy
Description
TET3 Polyclonal specifically detects TET3 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| TET3 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence | |
| EC 1.14.11.n2, hCG_40738, KIAA0401, methylcytosine dioxygenase TET3, MGC22014, probable methylcytosine dioxygenase TET3, tet oncogene family member 3 | |
| Produced in rabbits immunized with E. coli-derived Human TET3 fragment. | |
| 0.1 mL | |
| Epigenetics | |
| 200424 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunofluorescence | |
| FITC | |
| PBS | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction