missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TET3 Monoclonal Antibody (OTI10G4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803977S
This item is not returnable.
View return policy
Description
TET3 Monoclonal antibody specifically detects TET3 in Human samples. It is validated for Western BlotSpecifications
| TET3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O43151 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 200424 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI10G4 | |
| Unconjugated | |
| TET3 | |
| B430006D22Rik; BC037432; D230004J03Rik; hCG_40738; KIAA0401; Methylcytosine dioxygenase TET3; MGC22014; probable methylcytosine dioxygenase TET3; putative methylcytosine dioxygenase; Tet methylcytosine deoxygenase 3 isoform; tet methylcytosine dioxygenase 3; tet oncogene family member 3; Tet3 | |
| Human recombinant protein fragment corresponding to amino acids 241-568 of human TET3 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction