Learn More
TET3 Monoclonal Antibody (OTI11E11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA803972S
Encompass_Preferred
 View more versions of this product
Product Code. 17573273
  • € 242.00 / 30µL
Description
TET3 Monoclonal antibody specifically detects TET3 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot
Specifications
Specifications
TET3
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P08151
Mouse
Affinity chromatography
RUO
2735
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI11E11
Unconjugated
GLI1
B430006D22Rik; BC037432; D230004J03Rik; hCG_40738; KIAA0401; Methylcytosine dioxygenase TET3; MGC22014; probable methylcytosine dioxygenase TET3; putative methylcytosine dioxygenase; Tet methylcytosine deoxygenase 3 isoform; tet methylcytosine dioxygenase 3; tet oncogene family member 3; Tet3
Human recombinant protein fragment corresponding to amino acids 755-1106 of human GLI1 produced in E.coli.
30 μL