missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetanus Toxin Antibody, DyLight 594, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-63239DL594
This item is not returnable.
View return policy
Description
Tetanus Toxin Polyclonal antibody specifically detects Tetanus Toxin in Bacteria samples. It is validated for Western Blot, ELISA
Specifications
| Tetanus Toxin | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| DyLight 594 | |
| Tentoxylysin | |
| Purified tetanus toxin | |
| 0.1 mL | |
| Virology Bacteria and Parasites | |
| Bacteria | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction