missing translation for 'onlineSavingsMsg'
Learn More

(+)-Tetrabenazine, MedChemExpress

Product Code. p-200101491
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16488538

Brand: Medchem Express HYB0590B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (+)-Tetrabenazine ((+)-TBZ; (3R,11bR)-TBZ; (3R,11bR)-Tetrabenazine) is a reversible inhibitor of vesicular monoamine transporter 2 (VMAT-2), inhibits transport by VMAT2 with 10-fold greater potency than transport by VMAT1.

(+)-Tetrabenazine ((+)-TBZ

Specifications

Chemical Name or Material (+)-Tetrabenazine
CAS 1026016-83-0
Color White
Purity Grade Notes Research
Molecular Formula C19H27NO3
Quantity 10 mg
Synonym (+)-TBZ (3R,11bR)-TBZ (3R,11bR)-Tetrabenazine
Solubility Information DMSO : 16.67 mg/mL (52.52 mM; Need ultrasonic)
SMILES COC1=C(OC)C=C(CCN(C[C@H]2CC(C)C)[C@]3([H])CC2=O)C3=C1
Molecular Weight (g/mol) 317.42
Formula Weight 317.42
Percent Purity 95.64%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.