missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Tetracosactide

Product Code. 16493966
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16493966 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16493966 Supplier MedChemExpress Supplier No. HYP0060
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Tetracosactide (INN) is an analogue of adrenocorticotrophic hormone (ACTH), with the biological activity of stimulating production of corticosteroids in the adrenal cortex.

Specifications

Purity 98.04%
CAS 16960-16-0
Content And Storage Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
For Use With (Application) Cancer-programmed cell death
Molecular Weight (g/mol) 2933.44 g/mol
Product Type Peptides
Quantity 25 mg
Shipping Condition Room temperature
Solubility H2O : 100 mg/mL (34.09 mM∣Need ultrasonic)
Synonym Tetracosactrin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.