missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetradecanoic-12-d1 Acid, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
Specifications
Specifications
| Chemical Name or Material | Tetradecanoic-12-D Acid |
| CAS | 358730-97-9 |
| Molecular Formula | C14 D H27 O2 |
| Quantity | 0.25 g |
| Synonym | Tetradecanoic-12-d acid (9 CI), Myristic acid-12-d, Tetradecanoic acid-12-d, Tetradecanoic-12-D acid |
| InChI Formula | InChI=1 S/C14H28O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14(15)16/h2-13H2,1H3,(H,15,16)/i3D |
| SMILES | [2 H]C(CC)CCCCCCCCCCC(=O)O |
| IUPAC Name | 12-deuteriotetradecanoic acid |
| Molecular Weight (g/mol) | 229.377 |
| Formula Weight | 229.215 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction