missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylsilane, 97%, Thermo Scientific™

Catalog Number p-7077230
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
631-36-7
C8H20Si
144.34
VCZQFJFZMMALHB-UHFFFAOYSA-N
silane, tetraethyl, tetraethylsilicon, tetraethylsilane, acmc-209ndb, c2h5 4si
12426
tetraethylsilane
CC[Si](CC)(CC)CC
This item is not returnable. View return policy
631-36-7
C8H20Si
144.34
VCZQFJFZMMALHB-UHFFFAOYSA-N
silane, tetraethyl, tetraethylsilicon, tetraethylsilane, acmc-209ndb, c2h5 4si
12426
tetraethylsilane
CC[Si](CC)(CC)CC
Melting Point -82.5°C
Density 0.7600g/mL
Boiling Point 153.0°C to 154.0°C
Flash Point 26°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula Si(C2H5)4
Refractive Index 1.4250 to 1.4280
Quantity 5 g
Specific Gravity 0.76
Solubility Information Solubility in water: insoluble.
Formula Weight 144.34
Percent Purity 97%
Chemical Name or Material Tetraethylsilane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'