missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrahydro-N,N-dimethyl-2,2-diphenyl-3-furanmethanamine Hydrochloride (>75%), TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | Tetrahydro-N,N-dimethyl-2,2-diphenyl-3-furanmethanamine Hydrochloride |
| CAS | 195615-84-0 |
| Molecular Formula | C19H23NO . HCl |
| Quantity | 250 mg |
| Synonym | 3-Furanmethanamine, tetrahydro-N,N-dimethyl-2,2-diphenyl-, hydrochloride |
| InChI Formula | InChI=1S/C19H23NO.ClH/c1-20(2)15-18-13-14-21-19(18,16-9-5-3-6-10-16)17-11-7-4-8-12-17;/h3-12,18H,13-15H2,1-2H3;1H |
| SMILES | Cl.CN(C)CC1CCOC1(c2ccccc2)c3ccccc3 |
| IUPAC Name | 1-(2,2-diphenyloxolan-3-yl)-N,N-dimethylmethanamine;hydrochloride |
| Molecular Weight (g/mol) | 317.9 |
| Formula Weight | 317.1546 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction