missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrahydrocannabinol Antibody (TH2A), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07896C
This item is not returnable.
View return policy
Description
Tetrahydrocannabinol Monoclonal antibody specifically detects Tetrahydrocannabinol in Non-species specific samples. It is validated for ELISA
Specifications
| Tetrahydrocannabinol | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Non-species specific | |
| Purified |
| ELISA | |
| TH2A | |
| ELISA | |
| C21H30O2;Delta9-Tetrahydrocannabinol;Delta9-THC;Deltanyne;Dronabinol;Marinol;Tetrahydrocannabinol | |
| Tetrahydrocannabinol (THC) | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction