Learn More
Tetrahydrocannabinol Antibody (THC3), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-07897AF350
Product Code. 18511553
  • € 440.00 / 0.10mL
Description
Tetrahydrocannabinol Monoclonal antibody specifically detects Tetrahydrocannabinol in Non-species specific samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Tetrahydrocannabinol
Monoclonal
Alexa Fluor 350
50mM Sodium Borate
Mouse
Protein A purified
RUO
Non-species specific
Purified
ELISA
THC3
ELISA
C21H30O2;Delta9-Tetrahydrocannabinol;Delta9-THC;Deltanyne;Dronabinol;Marinol;Tetrahydrocannabinol
Tetrahydrocannabinol (THC)
0.1 mL
Primary
Store at 4°C in the dark.