missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrahydrocannabinol Antibody (THC3) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 323.00 - € 1050.00
Specifications
| Antigen | Tetrahydrocannabinol |
|---|---|
| Clone | THC3 |
| Concentration | 1 mg/mL |
| Applications | ELISA, Immunoassay |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18607827
|
Novus Biologicals
NBP3-07897 |
1 mg |
€ 1050.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18605076
|
Novus Biologicals
NBP3-07897-0.1mg |
0.1 mg |
€ 323.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tetrahydrocannabinol Monoclonal antibody specifically detects Tetrahydrocannabinol in Nonspecies specific samples. It is validated for ELISA, ImmunoassaySpecifications
| Tetrahydrocannabinol | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS | |
| Tetrahydrocannabinol (THC) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| THC3 | |
| ELISA, Immunoassay | |
| Unconjugated | |
| Mouse | |
| Non-species specific | |
| C21H30O2;Delta9-Tetrahydrocannabinol;Delta9-THC;Deltanyne;Dronabinol;Marinol;Tetrahydrocannabinol | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title