missing translation for 'onlineSavingsMsg'
Learn More

Tetrakis(dimethylamino)silane, 99%, Thermo Scientific Chemicals

Catalog Number p-7029608
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
1624-01-7
C8H24N4Si
204.39
MFCD00014859
SSCVMVQLICADPI-UHFFFAOYSA-N
tetrakis dimethylamino silane, silanetetramine, octamethyl, octamethylsilanetetramine, octamethylsilatetramine, silane, tetrakis dimethylamino, silane 48-12 tetrakis, dimethylamine, n,n',n,n'-silanetetrayltetrakis, silanetetramine, n,n,n',n',n,n,n',n'-octamethyl, n,n,n',n',n,n,n',n'-octamethylsilanetetraamine, si nme2 4
74194
N-methyl-N-[tris(dimethylamino)silyl]methanamine
CN(C)[Si](N(C)C)(N(C)C)N(C)C
This item is not returnable. View return policy
1624-01-7
C8H24N4Si
204.39
MFCD00014859
SSCVMVQLICADPI-UHFFFAOYSA-N
tetrakis dimethylamino silane, silanetetramine, octamethyl, octamethylsilanetetramine, octamethylsilatetramine, silane, tetrakis dimethylamino, silane 48-12 tetrakis, dimethylamine, n,n',n,n'-silanetetrayltetrakis, silanetetramine, n,n,n',n',n,n,n',n'-octamethyl, n,n,n',n',n,n,n',n'-octamethylsilanetetraamine, si nme2 4
74194
N-methyl-N-[tris(dimethylamino)silyl]methanamine
CN(C)[Si](N(C)C)(N(C)C)N(C)C

Silicon dioxide thin films prepared by chemical vapor deposition from tetrakis (dimethylamino) silane and ozone. Aminosilanes mere prepared by methods all essentially similar, and only the 15olation of tetrakis-(dimethylamino)-silane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Silicon dioxide thin films prepared by chemical vapor deposition from tetrakis (dimethylamino) silane and ozone. Aminosilanes mere prepared by methods all essentially similar, and only the 15olation of tetrakis-(dimethylamino)-silane.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
TRUSTED_SUSTAINABILITY
Boiling Point 196°C
Melting Point 15°C
Quantity 1 g
Linear Formula [(CH3)2N]4Si
UN Number UN3129
Flash Point 58°C (136°F)
Solubility Information Reacts with water.
Formula Weight 204.4
Percent Purity 99%
Odor Amine-like
Refractive Index 1.443
Sensitivity Moisture sensitive
Density 0.885
Chemical Name or Material Tetrakis(dimethylamino)silane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'