Learn More
Tetramethylsilane-d12, TRC
Product Code. 30426575
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30426575 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30426575 Supplier Toronto Research Chemicals Supplier No. TRCT30290950MG
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
Denna artikel kan inte returneras. Se returpolicy
Reglering: Begränsningar kan gälla för köp av denna produkt och vi kan behöva begära ytterligare dokumentation från dig.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Tetramethylsilane-D12
CAS 18145-38-5
Molecular Formula C4D12Si
Quantity 50 mg
Synonym Tetra(methyl-d3)silane, Tetrakis(trideuteromethyl)silane, Tetramethylsilane-d12
InChI Formula InChI=1S/C4H12Si/c1-5(2,3)4/h1-4H3/i1D3,2D3,3D3,4D3
SMILES [2H]C([2H])([2H])[Si](C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC Name tetrakis(methyl-d3)silane
Molecular Weight (g/mol) 100.3
Formula Weight 100.15