Learn More
Tetrapropylammonium Bromide, TRC
Product Code. 30449217
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30449217 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30449217 Supplier Toronto Research Chemicals Supplier No. TRCT310770500MG
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Tetrapropylammonium Bromide
CAS 1941-30-6
Molecular Formula C12H28BrN
Quantity 500 mg
Synonym 1-Propanaminium','N,N,N-tripropyl-','bromide (1:1) (ACI), 1-Propanaminium','N,N,N-tripropyl-','bromide (9CI), Ammonium','tetrapropyl-','bromide (8CI), Tetrapropylammonium bromide (6CI), Tetra-n-propylammonium bromide, TPABr
InChI Formula InChI=1S/C12H28N.BrH/c1-5-9-13(10-6-2,11-7-3)12-8-4;/h5-12H2,1-4H3;1H/q+1;/p-1
SMILES [Br-].CCC[N+](CCC)(CCC)CCC
IUPAC Name tetrapropylazanium;bromide
Molecular Weight (g/mol) 266.26
Formula Weight 265.1405