Learn More
TEV Protease Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP1-97669
Product Code. 18747153
  • € 691.00 / 0.10mg
Description
TEV Protease Polyclonal specifically detects TEV Protease in Virus samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polyclonal
Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.