Learn More
TEV Protease Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-97669
This item is not returnable.
View return policy
Description
TEV Protease Polyclonal specifically detects TEV Protease in Virus samples. It is validated for Western Blot, ELISA.
Specifications
| Polyclonal | |
| Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. | |
| Purified | |
| IgG |