missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TEX101 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560008
This item is not returnable.
View return policy
Description
Immunogen sequence: VTSYSNYCED SFCNDKDSLS QFWEFSETTA STVSTTLHCP TCVALGTCFS APSLPCPNGT TRCYQGKLEI TG Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 55%.
May play a role in signal transduction and promote protein tyrosine phosphorylation. Target protein is a glycoprotein whose aggregation induced a rapid increase in tyrosine phosphorylation of Syk kinase and LAT adaptor, calcium flux, and release of secretory components.
Specifications
| TEX101 | |
| Polyclonal | |
| Unconjugated | |
| TEX101 | |
| cancer/testis antigen 131; cell surface receptor NYD-SP8; CT131; GTPR867; NYD-SP8; PRO1884; Scleroderma-associated autoantigen; SGRG; SPATA44; spermatogenesis associated 44; spermatogenesis-related gene protein; TES101RP; testis expressed 101; testis expressed sequence 101; testis-expressed protein 101; testis-expressed sequence 101 protein; testis-specific protein TES101RP; TEX101; UNQ867/PRO1884 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 83639 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BY14 | |
| TEX101 | |
| Recombinant protein corresponding to Human TEX101. Recombinant protein control fragment (Product #RP-99088). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction