missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TEX101 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120611
This item is not returnable.
View return policy
Description
Positive test controls include: A-549. Immunogen sequence: LYCQKGLSMT VEADPANMFN WTTEEVETCD KGALCQETIL IIKAGTETAI LATKGCIPEG EEAITIVQHS SPPGLIVTSY SNYCEDSFCN DKDSLSQFWE FSETTAS.
May play a role in signal transduction and promote protein tyrosine phosphorylation. Target protein is a glycoprotein whose aggregation induced a rapid increase in tyrosine phosphorylation of Syk kinase and LAT adaptor, calcium flux, and release of secretory components.
Specifications
| TEX101 | |
| Polyclonal | |
| Unconjugated | |
| TEX101 | |
| cancer/testis antigen 131; cell surface receptor NYD-SP8; CT131; GTPR867; NYD-SP8; PRO1884; Scleroderma-associated autoantigen; SGRG; SPATA44; spermatogenesis associated 44; spermatogenesis-related gene protein; TES101RP; testis expressed 101; testis expressed sequence 101; testis-expressed protein 101; testis-expressed sequence 101 protein; testis-specific protein TES101RP; TEX101; UNQ867/PRO1884 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 207113, 56746, 83639 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1.4 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q924B5, Q9BY14, Q9JMI7 | |
| TEX101 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 44-150 of human TEX101 (NP_113639.4). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction